missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals

Product Code. 15439368
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247570-24-7
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD03844604
InChI Key FEYLUKDSKVSMSZ-UHFFFAOYSA-N
Synonym trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine
PubChem CID 2756050
IUPAC Name tert-butyl N-(4-aminocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)N

Specifications

Quantity 1 g
Solubility Information Soluble in methanol,ethyl acetate and Slightly soluble in water.
Formula Weight 214.3
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.