missing translation for 'onlineSavingsMsg'
Learn More
cis-3-Octen-1-ol, 95%, Thermo Scientific Chemicals Product Code.: 11409326

cis-3-Octen-1-ol, 95%, Thermo Scientific Chemicals

Product Code. 11409326
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11409326

Brand: Thermo Scientific Alfa Aesar A19014.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20125-84-2
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014062
InChI Key YDXQPTHHAPCTPP-WAYWQWQTSA-N
Synonym cis-3-octen-1-ol, z-3-octen-1-ol, z-oct-3-en-1-ol, 3-octen-1-ol, z, unii-34a1x2y8m9, 3z-octen-1-ol, 3z-3-octen-1-ol, 3-octenol, cis, fema no. 3467, 3-octen-1-ol, 3z
PubChem CID 5364519
IUPAC Name (Z)-oct-3-en-1-ol
SMILES CCCCC=CCCO

Specifications

Density 0.85
Boiling Point 88°C to 89°C (15 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.447
Quantity 50 g
Beilstein 1720075
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material cis-3-Octen-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP02099 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19014.18,A19014.09
Lot Number10232374 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19014.18,A19014.09
Lot Number10167641 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19014.18,A19014.09
Lot Number9002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19014.18,A19014.09
Lot Number10215478 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19014.18,A19014.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-3-Octen-1-ol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.