missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%

Product Code. 10607484 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
This item is not returnable. View return policy

Product Code. 10607484

Brand: Thermo Scientific Acros 277740020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 37910-65-9 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37910-65-9
MDL Number MFCD00143982
InChI Key JWYOAMOZLZXDER-UHNVWZDZSA-N
Synonym 1r,2s-2-aminocyclopentanecarboxylic acid, cispentacin, cis-2-amino-1-cyclopentanecarboxylic acid, --cispentacin, cis-pentacin, antibiotic fr 109615, 1r,2s-2-aminocyclopentane-1-carboxylic acid, --1r,2s-cispentacin, +/--cispentacin, cispentacin,-
PubChem CID 73305
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxylic acid
SMILES C1CC(C(C1)N)C(=O)O

Specifications

Melting Point 218.0°C to 220.0°C
Assay Percent Range 98+%
Molecular Formula C6H11NO2
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Molecular Weight (g/mol) 129.16
Quantity 2 g
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.