missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(Vi) Oxide, puriss. p.a., ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 50 g |
UN Number | 1463 |
Formula Weight | 99.99g/mol |
Percent Purity | ≥99% (RT) |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Chromium(Vi) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'