missing translation for 'onlineSavingsMsg'
Learn More

Chromium(Vi) Oxide, puriss. p.a., ACS Reagent, Honeywell Fluka™

Product Code. 15642880
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Quantity 250 g
UN Number 1463
Formula Weight 99.99g/mol
Percent Purity ≥99% (RT)
Grade ACS/puriss. p.a.
Packaging Glass Bottle
Chemical Name or Material Chromium(Vi) Oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'