Learn More
Bochem™ Chromium-Plated Burette Clamp
Brand: Bochem™ 5623
This item is not returnable.
View return policy
Specifications
130 mm | |
Shaft | |
10 mm |
Chromium-plated | |
Buret Clamp |
Product Content Correction
Product Title
Bochem™ Chromium-Plated Burette Clamp > Dia.: 10 mm