Learn More
Fisherbrand™ Chromatography Columns
Increase sample throughput and laboratory productivity with Fisherbrand™ Chromatography Columns for all compound separation needs.
Brand:  Fisherbrand™ R2/10/3
 View more versions of this product
Product Code. 15379064
  • € 84.75 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Description
  • For separation of compounds
  • With sintered glass fritted disk
  • With PTFE keys
Specifications
Specifications
1
Chromatography Column
3
100 mm
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Chromatography Columns > Porosity: 3