Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Brand: Macherey-Nagel™ 730356
This item is not returnable.
View return policy
Description
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
Polypropylene Columns | |
45 mL | |
250 Pack |
Frit | |
Polyethylene |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Polyethylene Frit > Volume: 45mL