Learn More
Macherey-Nagel™ Chromabond™ Glass Empty SPE Column
Brand: Macherey-Nagel™ 730172
This item is not returnable.
View return policy
Specifications
6 mL | |
Solid Phase Extraction | |
Glass fiber frits, filter element |
Empty | |
Empty Solid Phase Extraction Column |
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Glass Empty SPE Column > Column Capacity: 6mL