Learn More
Minigrip™ Resealable Write-on Bags
Click to view available options
Width (Metric):
40 mm
75 mm
100 mm
140 mm
190 mm
Height (Metric):
65 mm
85 mm
140 mm
190 mm
missing translation for 'unitSize'
Pack of 100
Pack of 1000
Description
- Polyethylene
- Resealable
- Autoclavable
- Three write-on bars for labelling
Specifications
Specifications
| Description | Resealable Bag |
| Color | Clear |
| For Use With (Application) | Ideal for storing stationery and sending samples |
| Material | Polyethylene |
| Quantity | 100 Pack |
| Width (Metric) | 75 mm |
| Height (Metric) | 85 mm |