Learn More
Emma Safety Footwear Chicago Safety Shoes
Light and comfortable with knitted textile upper and Nano Carbon Composite protective nose. The TPU GripForce™ Easy Twist outsole offers maximum grip, while The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
Brand: Emma Safety Footwear MM40264740
Description
- S1P-Safety shoe with sole plate for dry work
- Knitted Textile construction
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD Certified
Specifications
EN ISO 20345:2011 | |
Gray, Black | |
Hydro-Tec™ Antibacterial | |
40 |
Laces | |
Chicago | |
Textile | |
Composite |
Product Suggestions
Customers who viewed this item also viewed