Learn More
Emma Safety Footwear Chicago Safety Shoes
Light and comfortable with knitted textile upper and Nano Carbon Composite protective nose. The TPU GripForce™ Easy Twist outsole offers maximum grip, while The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
Brand:  Emma Safety Footwear MM40264746
 View more versions of this product
Product Code. 15938401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile construction
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD Certified
Specifications
Specifications
EN ISO 20345:2011
Gray, Black
Hydro-Tec™ Antibacterial
46
Laces
Chicago
Textile
Composite
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Chicago Safety Shoes > Size: 46