Learn More
Liebherr™ Chest Freezer
Product Code. 12014672 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
299 L
485 L
601 L
Unit Size:
Each
Product Code. 12014672
Brand: Liebherr™ GTL 6105
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ Chest Freezers feature aluminum interior containers and stainless steel adjustable feet.
  • Steel chest freezer
  • Mechanic control system
  • External Analog display
  • Ergonomic handle with aluminum trim
  • Fitted lock
Specifications
Capacity (Metric) 601 L
Temperature Range -14°C to -26°C
Color White
Model GTL6105
Dimensions (L x W x H) Exterior 91.7 H x 164.7 W x 77.6 D cm
Depth (Metric) Interior 57.5 cm
Height (Metric) Interior 70.2 cm
Width (Metric) Interior 152 cm
Voltage 220/240 V
Frequency 50 Hz
Refrigerant R600A