missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics CHEMets Phenol Visual Water Testing Kit

Product Code. 11791004
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 11791004

Brand: CHEMetrics K8012

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

REMOVE

Provides results in two minutes or less

Provide results in two minutes or less

Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, 25 mL sample cup and instructions

Specifications

Analytes Phenol
Includes Plastic case, Refill, Low and High Range Comparators, 25 mL sample cup and instructions
No. of Tests 30
Range 0 to 1 ppm and 0 to 12 ppm
Test Method 4-Aminoantipyrine
Test Time 1 min.
Type CHEMets Visual Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • Phenols
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H400-Very toxic to aquatic life.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.