Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR
Product Code. 11768999
Click to view available options
Range:
20 to 800mg/L
5 to 500mg/L
Unit Size:
Each
Product Code. 11768999
Brand: Lovibond™ 414240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accurate test kit for routine water analysis
Specifications
For Use With (Application) Routine water analysis
Includes 30mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case
No. of Tests 100
Range 5 to 500mg/L
Type Total Hardness Mini Kit