Learn More
Honeywell™ Check and Go DeepTril 1 Gloves
Click to view available options
Size:
7
8
9
10
Description
- Black polyamide knitted glove
- Fully dipped black foam nitrile coating with PVC dots on the palm
- The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The nitrile foam and PVC dots on palm improves the grip and optimizes the glove's breathability
- The knitted and elasticated wrist gives the glove good support
- The nitrile coating provides outstanding abrasion resistance and impermeability to oils
- The black color is suitable for work in a soiled atmosphere
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2003 Mechanical hazards |
Description | 13 gauge |
Color | Black |
Material | Foam Nitrile |
Size | 10 |
Length (Metric) | 25 cm |