Learn More
Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures
Compatible with automated systems. Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures are used for anchorage dependant cell lines.
€ 256.25 - € 307.50
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Material | Polystyrene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10070282
|
Greiner Bio-One
657185 |
16 mL |
€ 256.25
Pack of 100 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
10156610
|
Greiner Bio-One
662102 |
3.3 mL (Well) |
€ 256.25
Pack of 100 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
10345791
|
Greiner Bio-One
665102 |
6.5 mL |
€ 307.50
Pack of 100 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
10632672
|
Greiner Bio-One
677102 |
1.7 mL |
€ 274.67
Pack of 100 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
- Manufactured from transparent virgin polystyrene
- Low auto fluorescence
- Single position lids to prevent cross-contamination
- Alphanumeric well coding
- With hydrophobic surface for suspension cultures and hybridoma cell
Service Plan
No
Warranty
No
Specifications
Clear | |
Polystyrene | |
Polystyrene | |
Sterile | |
Lid | |
1 | |
127.76 mm | |
19 mm |
Closure Included | |
Flat | |
Rectangular | |
Multi-well Cell Culture Plate | |
100 | |
127.8 x 85.5 x16.5 mm | |
85.48 mm |
Product Title
Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures