Learn More
Molecular Probes™ CellROX™ Orange Flow Cytometry Assay Kit
CellROX™ Orange Flow Cytometry Assay Kit
Brand: Molecular Probes™ C10493
317.10 EUR valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The CellROX™ Orange Flow Cytometry Assay Kit enables the flow cytometric detection of reactive oxygen species (ROS) in live cells. The kit includes the novel, fluorogenic CellROX™ Orange Reagent, as well as SYTOX™ Red Dead Cell Stain, N-acetyl cysteine (an antioxidant, for negative control), and tert-butyl hydroperoxide solution (TBHP, an inducer of ROS).View a selection guide for all CellROX™ Reagents and Kits.
The CellROX™ Orange Flow Cytometry Assay Kit features:
The CellROX™ Orange Detection Reagent is cell-permeable and non-fluorescent or very weakly fluorescent while in the reduced state. Upon oxidation, the reagent exhibits a strong fluorogenic signal that has absorption/emission maxima of 545/565 nm and remains localized in the cytoplasm. When used together with the included SYTOX™ Red Dead Cell Stain, oxidatively stressed and nonstressed cells are reliably distinguished from dead cells by flow cytometry.
Specifications
Tube | |
100 assays | |
CellROX orange: 545/565, SYTOX red: 640/658 |