Learn More
Greiner Bio-One CELLMASTER™ Glycolised Polyethylene Terephthalate (PETG) 2X Roller Bottles - Short Form
Click to view available options
Capacity (Metric):
2500 mL
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic, non-cytotoxic
- Seamless blow-moulding technique, rules out leaking seams
- USP Class VI end product testing certified
- Safety screw cap guarantees tightly closed, co
Specifications
Specifications
Bottle Surface | Ribbed |
Capacity (Metric) | 2500 mL |
Color | Clear |
Diameter (English) | 4.8 in. |
For Use With (Application) | Cell cuture |
Material | Polystyrene |
No. per Case | 5 |
Sterility | Sterile |
Cell Growth Area | 1700 cm2 |
Certified | USP Class VI end product testing |