missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD8 Fab-TACS™ Gravity Kit

IBA Lifesciences™ CD8 Fab-TACS™ Gravity Kit is for reversible cell isolation delivering label-free target cells.

Brand:  IBA Lifesciences 6-3203-010

 View more versions of this product

Product Code. 15899784

This item is not returnable. View return policy

Description

Description

  • The kit allows the selection of CD8+ T cells directly from whole blood.
  • CD8 Fab-Strep, human
  • Buffer CI (10x)
  • Biotin stock solution
  • Fab-TACS™ Gravity Adapter
  • Fab-TACS™ Flow Restrictor
  • The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
  • Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
  • Subsequently, whole blood or other blood preparations pass through the column.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
  • Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
  • After elution, the Fab-Streps self-dissociate from the cell surfaces.
  • The label-free authentic target cells are now ready for further downstream applications.
Specifications

Specifications

Non-magnetic Cell Isolation Kit
Human Whole Blood
500 mL whole blood
2°C to 8°C
Isolation of label-free cells
10 Columns, Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IBA Lifesciences CD8 Fab-TACS™ Gravity Kit > Quantity: 10 Columns, Human

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.