missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD8 Fab-TACS™ Gravity Kit
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
10 Columns, Human
4 Columns, Human
Unit Size:
1 set
Description
- The kit allows the selection of CD8+ T cells directly from whole blood.
- CD8 Fab-Strep, human
- Buffer CI (10x)
- Biotin stock solution
- Fab-TACS™ Gravity Adapter
- Fab-TACS™ Flow Restrictor
- The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
- Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
- Subsequently, whole blood or other blood preparations pass through the column.
- Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
- Non-target cells are efficiently washed away.
- In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
- After elution, the Fab-Streps self-dissociate from the cell surfaces.
- The label-free authentic target cells are now ready for further downstream applications.
Specifications
Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| Donor Source | Human Whole Blood |
| For Use With (Application) | Isolation of label-free cells |
| Sufficient For | 200 mL whole blood |
| Quantity | 4 Columns, Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction