missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 319.00 - € 648.00
Specifications
| Antigen | CD63 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620625
|
Novus Biologicals
NBP3-21363-25ul |
25 μL |
€ 319.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687644
|
Novus Biologicals
NBP3-21363-100ul |
100 μL |
€ 648.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD63 Polyclonal antibody specifically detects CD63 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| CD63 | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| PBS, pH 7.2, 40% glycerol | |
| 967 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VGAQLVLSQTIIQGATPGSL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title