Learn More
IBA Lifesciences CD45RA Isolation Kit for FABian™
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit, Human
10 Kits, Human
Unit Size:
1 set
10 sets
Description
Specifications
Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| Donor Source | Human Whole Blood or Buffy Coat |
| For Use With (Application) | Isolation of label-free cells |
| Sufficient For | 9 mL Whole Blood |
| Quantity | 10 Kits, Human |