missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD144 Rat anti-Mouse, BB700, Clone: 11D4.1, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 742179
This item is not returnable.
View return policy
Description
CD144 Monoclonal antibody specifically detects CD144 in Mouse samples. It is validated for Flow CytometrySpecifications
CD144 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
Cdh5; Cadherin-5; CADH5; VE-cadherin; Vascular endothelial cadherin; 7B4 | |
Mouse VE-Cadherin-Ig Fusion | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
11D4.1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BB700 under optimal conditions that minimize unconjugated dye and antibody. |