Learn More
Applied Biosystems™ Cathode Buffer Container (CBC), for 3500/SeqStudio™ Flex
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.
Brand: Applied Biosystems™ 4408256
174.00 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The container has two separate compartments, the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections.The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
Order Info
4 Pack
Specifications
Contains 4 packs of cathode buffer container. Store the Cathode Buffer Container at 2-8°C until ready to use. Refer to the expiration date on the label. | |
Cathode Buffer Container | |
4 pack |
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers | |
Sustainable packaging |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2411047 | Certificate of Analysis | 23/01/2025 | 4408256 |
2411046 | Certificate of Analysis | 20/01/2025 | 4408256 |
2410045 | Certificate of Analysis | 15/01/2025 | 4408256 |
2410044 | Certificate of Analysis | 09/01/2025 | 4408256 |
2410043 | Certificate of Analysis | 27/12/2024 | 4408256 |