Learn More
Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs
Code produit. 16384056 Tous les produits Epredia Produits
missing translation for 'orderingAttributeHoverText'
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
missing translation for 'unitSize'
Pack of 750
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 16384056
missing translation for 'mfr': Epredia™ B851729OR
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Spécification
Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Front Hinge
PrintMate Compatible Yes
Color Orange
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette II Biopsy Cassette