missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase III Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA550993

 View more versions of this product

Product Code. 17918782

  • € 379.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase III
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
P07451, P14141, P16015, Q5S1S4
Ca3, Car3
Recombinant Human CA3 protein, Ala2-Lys260 (Accession #P07451)
200 μL
Primary
Human, Mouse, Porcine, Rat
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
C2
Unconjugated
Ca3
BB219044, CA III, Ca3, CAH3, CAIII, CA-III, CAIII (Muscle), Car3, Car-3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, carbonic anhydrase-like protein, EC 4.2.1.1, epididymis secretory sperm binding protein Li 167mP, HEL-S-167mP
Mouse
Protein A/G
RUO
12350, 494016, 54232, 761
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers