Learn More
Erlab™ Captair Smart Stainless Steel Work Surface
Product Code. 15943940
Click to view available options
For Use With (Equipment):
Smart 392 Fume Hood
Smart 481 Fume Hood
Unit Size:
Each
Product Code. 15943940
Brand: Erlab™ I0163050001
Erlab™ Captair Smart Stainless Steel Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
  • Constructed of stainless steel
  • For use with Captair Smart Ductless Filtering Fume Hoods
Specifications
For Use With (Equipment) Smart 392 Fume Hood
Includes Spill Tray
Depth (Metric) 648 mm
Width (Metric) 855 mm
Product Type Work Surface
Material Stainless Steel (304)