Learn More
Erlab™ Captair Smart Glass Work Surface with Retention Tray
Click to view available options
For Use With (Equipment):
Smart 321 Fume Hood
Smart 391 Fume Hood
Smart 392 Fume Hood
Smart 481 Fume Hood
Smart 483 Fume Hood
Smart 633 Fume Hood
Smart 714 Fume Hood
Unit Size:
Each
Description
- Tempered enameled glass worktop
- Includes retention tray filtering fume hoods
- Jointed frame
- Ergonomic arm rest for a comfortable working position
Specifications
Specifications
| For Use With (Equipment) | Smart 633 Fume Hood |
| Includes | Retention Tray |
| Depth (Metric) | 585 mm |
| Description | Jointed Frame |
| Width (Metric) | 1503 mm |
| Product Type | Work Surface |
| Material | Tempered Glass |