Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand:  Hirschmann™ 9212375
 View more versions of this product
Product Code. 10223341
  • € 140.00 / Pack of 1000
Explore more special offers
Description
Description
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Melting Point Determination Tube, One End Closed
1.55 mm
75 mm
1 x 1000/Pk.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed > Length Tube: 75 mm, Diameter Inner: 1.55 mm, Quantity: 1 x 1000/Pack