Learn More
Hirschmann™ Capillary Tubes, Both Ends Open
Product Code. 10709581
Click to view available options
Length (Metric) Tube:
60 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Inner:
0.95 mm
1.15 mm
1.30 mm
1.55 mm
Unit Size:
Pack of 1000
Product Code. 10709581
Brand: Hirschmann™ 9202312
Excellent for working with small samples in the laboratory. Can be utilized for a wide range of scientific and medical applications.
  • Both ends open
  • Available in a wide range of diameters/lengths
Specifications
Product Type Capillary Tubes, Both Ends Open
Diameter (Metric) Inner 1.55 mm
Length (Metric) Tube 120 mm
Quantity 4 x 250/Pk.