Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 10253671
Click to view available options
Length (Metric):
10 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Outer:
1 mm
1.35 mm
1.55 mm
1.75 mm
2.35 mm
Unit Size:
Pack of 1000
Product Code. 10253671
Brand: Hirschmann™ 9211712
Only null left
Add to basket
Add to basket
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Diameter (Metric) Inner 1.3 mm
Diameter (Metric) Outer 1.75 mm
For Use With (Application) Melting point determination
Tube Style One end closed
Length (Metric) 120 mm
Type Capillary Tube