Learn More
behr Labor-Technik™ HDPE, Canister with Screw Cap and UN Approval
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
25 L
30 L
60 L
Description
- White High-Density Polyethylene canisters
- UN-certified
- Screw-on cap
- Suitable for hazardous goods
Specifications
Specifications
Capacity (Metric) | 5 L |
Certifications/Compliance | UN-certified |
Closure Type | Screw Cap |
Color | White |
Description | Kanister 5 l, PE/natur |
Includes | Screw cap |
Material | HDPE |
Product Type | Canister |
Weight (Metric) | 280 g |
Product Content Correction
Product Title
behr Labor-Technik™ HDPE, Canister with Screw Cap and UN Approval >