Learn More
Candesartan-d4, MedChemExpress
MedChemExpress Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.
Brand: Medchem Express HY-B0205S
This item is not returnable.
View return policy
Description
Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.Specifications
Candesartan-d4 | |
10 mg | |
C24H16D4N6O3 | |
CV-11974-d4 | |
[2H]C1=C(CN2C(C(C(O)=O)=CC=C3)=C3N=C2OCC)C([2H])=C([2H])C(C4=CC=CC=C4C5=NNN=N5)=C1[2H] | |
444.48 | |
Research |
1346604-70-3 | |
Research | |
Solid | |
White | |
444.48 | |
98.85% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Candesartan-d4, MedChemExpress
Research purposes only