Learn More
Calsequestrin 2 Mouse anti-Human, CoraLite 594, Clone: 1C10A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66419-100UL
Description
The Calsequestrin 2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human Calsequestrin 2. This antibody recognizes human, rat, pig, mouse antigen. The Calsequestrin 2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| Calsequestrin 2 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 845 | |
| IgG2a |
| Immunofluorescence | |
| 1C10A1 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |