Learn More
BINDER Calibration Certificate Oxygen
Shop All BINDER Products
Click to view available options
Calibration Points:
1% Volume by Oxygen
80% Oxygen by Volume
Unit Size:
Each
Description
O2 Calibration certificate for carbon dioxide incubator versions with O2 control.
- Measuring point in center of chamber at 1 volume
Specifications
Specifications
| For Use With (Equipment) | CB/CBF Carbon Dioxide Incubators |
| Calibration Points | 1% Volume by Oxygen |
| Duration | 1 Day |
| Calibration Type | Center of Chamber |
| Product Type | Calibration Certificate |
| Service Offering | Center of Chamber Calibration |