Learn More
Calibration Buffer Solutions, Hanna Instruments™
Click to view available options
Quantity:
20 mL
1 L
pH:
4.01
9.18
10.01
Unit Size:
1000mL
Pack of 25
Description
- Premium quality pH 10.01 calibration buffer solution
- 25 sachets containing 20 mL of buffer
- Color coded label for easy pH buffer value identification
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- In a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Specifications
| Color | Colorless |
| pH | 9.18 |
| Physical Form | Liquid |
| Quantity | 20 mL |
| Solution Type | Buffer |
| Formulation | Aqueous Buffer Solution |
| Odor | Odorless |