missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Specifications
Specifications
Color | White |
pH | 7 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 172.17g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'