missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 1 L |
IUPAC Name | calcium;dichloride |
Formula Weight | 110.99 |
Concentration or Composition (by Analyte or Components) | 100mM aq. soln. |
Chemical Name or Material | Calcium chloride, sterile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'