missing translation for 'onlineSavingsMsg'
Learn More

Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles

Product Code. 16448213
Click to view available options
Diameter (English) Ball:
0.074 in.
0.078 in.
Gauge:
18G
20G
Length (English):
1.2 in.
1.5 in.
2 in.
3 in.
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 16448213

Brand: Cadence Science™ 9924

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For critical applications requiring sterile, flexible, single-use feeding needles

  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box

Specifications

Product Type Soft, Flexible Animal Feeding Needle
Material PTFE
Disposable Single-use
For Use With (Application) For critical applications requiring sterile, flexible, single use feeding needles
Gauge 18G
Sterility Sterile
Length (English) 3 in.
Length (Metric) 0.75 cm
Diameter (English) Ball 0.078 in.
Diameter (Metric) Ball 0.02 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.