Learn More
Uvex™ C300 Dry Cut Protection Gloves
Product Code. p-7236911 Shop All Uvex Products
Click to view available options
Size:
8
Unit Size:
Each
Product Code. 15357102
Brand: Uvex™ 6054908
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Uvex C300 Dry gloves are designed to provide level 3/C cut protection and feature HPV dots on the palm for enhanced grip capabilities.
  • Gloves provide level 3/C blade cut resistance, and very high tear resistance
  • High-performance vinyl (HPV) dots on the palm enhance grip capabilities in dry environments
  • Uvex Climazone technology minimizes excessive warmth and moisture
  • Highly flexible with very good tactile sensation
  • Silicone-free according to the imprint test
  • 3D Ergo technology ensures a secure fit
TRUSTED_SUSTAINABILITY
Specifications
Coating Material HPV Dots
Color Gray, Green
Material Bamboo Viscose, HPPE, Polyamide, Glass
Size 8
EN388 Standard XX4XC
Glove Cuff Style Knit
Series C300 Dry