Learn More
C14orf106 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26264-1-AP-20UL
 View more versions of this product
Product Code. 16864775
  • € 145.00 / 20µL
Explore more special offers
Description
Description
MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.
Specifications
Specifications
C14orf106
Polyclonal
Unconjugated
MIS18BP1
C14orf106, HSA242977, HsKNL 2, KIAA1903, KNL2, M18BP1, Mis18 binding protein 1, MIS18BP1, P243
Rabbit
Antigen Affinity Chromatography
RUO
55320
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P0N0
MIS18BP1
C14orf106 Fusion Protein Ag23483
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers