Learn More
Wheaton™ C Model 1R2P 2.0 Roller Culture Apparatus
Shop All Wheaton Science Products Products
Click to view available options
No. of Positions:
15
Unit Size:
Each
Specifications
Specifications
| Speed Range | 0.25 to 8.1 rpm |
| Electrical Requirements | 100/240 V, 50/60 Hz, 35 W |
| Voltage | 100/240 V |
| Temperature Range (Metric) | 10°C to 40°C |
| Wattage | 35 W |
| Certifications/Compliance | CE, CSA, UL |
| Frequency | 50/60 Hz |
| Depth (Metric) | 63 cm |
| Height (Metric) | 65 cm |
| Width (Metric) | 79 cm |
| Show More |