Learn More
Abeba™ Business Men 33240 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand:  Abeba™ 33240/41
 View more versions of this product
Product Code. 15609827
  • € 174.00 / 1 pair
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Microfiber
41
Rubber
Steel
Black
Leather
Slip-resistant
33240