Learn More
Abeba™ Business Men 33230 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand: Abeba™ 33230/38
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 S2 SRA
- Smooth leather
- Black
- Steel toe cap
- Slip resistance special rubber outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Microfiber | |
38 | |
Rubber | |
Steel |
Black | |
Leather | |
Slip-resistant | |
33230 |