Learn More
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ Product Code.: 10665492
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Product Code. 10665492
2.5 L, Plastic container
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Plastic container
Product Code. 10665492
Brand: Fisher Chemical J/2810/17
Request Bulk or Custom Format
Chemical Identifiers
MDL Number 217672
Specifications
Color Colorless
pH 3
Physical Form Liquid
Quantity 2.5 L
Packaging Plastic container
Product Content Correction
Product Title
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ >