missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.64, METTLER TOLEDO™

Product Code. p-8014385 Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 10598471

Brand: Mettler Toledo 51319013

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Specifications

Color Colorless
pH 4.64
Physical Form Liquid
Quantity 250 mL
Solution Type pH Buffer
Odor None
Packaging 250mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.