missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 13, Chem Lab™
Click to view available options
Quantity:
1 L
Specifications
Specifications
Melting Point | 0°C |
Color | Colorless |
pH | 13 |
Physical Form | Liquid |
Quantity | 1 L |
Solution Type | Buffer |
Formulation | Glycine-NaCl-NaOH |
Odor | Odorless |
Solubility Information | Water Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction