missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 11, Chem Lab™

Product Code. 10213962
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 0°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Formulation 70g NH4Cl + 570mL NH4OH 25%/H2O
Odor Ammonia
Solubility Information Water Soluble
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium chloride-Ammonia buffer pH 11
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium chloride, ammonium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'