Learn More
Buffer Solution pH 1.68, Reagecon™ Product Code.: 10276657
Buffer Solution pH 1.68, Reagecon™
Product Code. 10276657 Shop All Reagecon Products
500 mL
Click to view available options
Quantity:
500 mL
Product Code. 10276657
Brand: Reagecon EP168
Specifications
pH 1.68
Quantity 500 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number167724B1 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog NumberEP168
Lot Number167723H1 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog NumberEP168
Lot Number167722M1 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog NumberEP168
Lot Number167722F1 Certificate TypeCertificate of Analysis Date08/12/2022 Catalog NumberEP168
Lot Number167721G1 Certificate TypeCertificate of Analysis Date30/11/2021 Catalog NumberEP168
Product Content Correction
Product Title
Buffer Solution pH 1.68, Reagecon™ >